Assay Details
Target Gene Details
Entrez Gene ID: | 11234 |
Gene Name: | HPS5, biogenesis of lysosomal organelles complex 2 subunit 2 |
Gene Aliases: |
AIBP63, BLOC2S2 |
Location: |
Chr.11:18278670-18322498 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
More Information
Set Membership: |
|

Genomic Map