Assay Details
Target Gene Details
Entrez Gene ID: | 6691 |
Gene Name: | serine peptidase inhibitor, Kazal type 2 |
Gene Aliases: |
HUSI-II |
Location: |
Chr.4:56809860-56822210 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK2 | NM_001271718.1 | 4 | 633 | NP_001258647.1 |
| NM_001271719.1 | 4 | 593 | NP_001258648.1 | |
| NM_001271720.1 | 4 | 588 | NP_001258649.1 | |
| NM_001271721.1 | 4 | 548 | NP_001258650.1 | |
| NM_001271722.1 | 2 | 479 | NP_001258651.1 | |
| NM_021114.3 | 4 | 610 | NP_066937.1 | |
| NR_073417.1 | 4 | 462 | ||
| NR_073418.1 | 3 | 500 | ||
| NR_073419.1 | 4 | 502 | ||
| XM_011534405.1 | 3 | 724 | XP_011532707.1 | |
| AA776754.1 | 1 | 69 | ||
| AA973772.1 | 1 | 154 | ||
| AV743457.1 | 3 | 394 | ||
| BC022514.1 | 4 | 472 | AAH22514.1 | |
| BF589674.1 | 1 | 134 | ||
| BI520020.1 | 4 | 522 | ||
| BI561149.1 | 4 | 635 | ||
| BI562269.1 | 4 | 587 | ||
| BU853655.1 | 4 | 466 | ||
| DB451710.1 | ||||
| GU727622.1 | 4 | 472 | ||
| HY020998.1 | ||||
| X57655.1 | 4 | 471 | CAB37834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5214n100 | Chr.4:56786612 - 56999637 on Build GRCh38 | Gain |
|
| nsv1161088 | Chr.4:56678187 - 56838222 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map