Assay Details
Target Gene Details
Entrez Gene ID: | 115948 |
Gene Name: | coiled-coil domain containing 151 |
Gene Aliases: |
CILD30 |
Location: |
Chr.19:11420605-11435782 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC151 | NM_001302453.1 | 8 | 910 | NP_001289382.1 |
| NM_001302454.1 | 6 | 1024 | NP_001289383.1 | |
| NM_145045.4 | 8 | 1204 | NP_659482.3 | |
| XM_017026241.1 | 8 | 1143 | XP_016881730.1 | |
| AK056363.1 | 8 | 1125 | ||
| AK294355.1 | 10 | 1104 | ||
| AK302113.1 | 8 | 911 | ||
| AK308072.1 | 6 | 986 | ||
| BC014252.2 | 7 | 761 | AAH14252.2 | |
| BC141828.1 | 8 | 1136 | ||
| BC142637.1 | 8 | 1149 | ||
| BC144421.1 | 6 | 683 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833752 | Chr.19:11208324 - 11448939 on Build GRCh38 | Loss |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
| nsv953975 | Chr.19:11412925 - 11424131 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map