Assay Details
Target Gene Details
Entrez Gene ID: | 9846 |
Gene Name: | GRB2 associated binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.11:78215290-78417822 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAB2 | NM_012296.3 | 10 | 2465 | NP_036428.1 |
| NM_080491.2 | 10 | 2521 | NP_536739.1 | |
| XM_006718753.2 | 11 | 2566 | XP_006718816.1 | |
| XM_011545408.2 | 9 | 2172 | XP_011543710.1 | |
| AB011143.1 | 10 | 2465 | ||
| AB018413.1 | 10 | 2419 | BAA76737.1 | |
| BC152459.1 | 10 | 2465 |
Target Gene Details
Entrez Gene ID: | 57558 |
Gene Name: | ubiquitin specific peptidase 35 |
Gene Aliases: |
- |
Location: |
Chr.11:78188812-78233651 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP35 | XM_017018052.1 | XP_016873541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv527066 | Chr.11:78172809 - 78252087 on Build GRCh38 | Gain |
|
| nsv1036793 | Chr.11:78198402 - 78265821 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map