Assay Details
Target Gene Details
Entrez Gene ID: | 345557 |
Gene Name: | phosphatidylinositol specific phospholipase C X domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.5:41306946-41510628 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCXD3 | NM_001005473.2 | 3 | 7605 | NP_001005473.1 |
| AK124638.1 | 2 | 3741 | ||
| BX648329.1 | 4 | 7416 | CAH56150.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604840 | Chr.5:41261866 - 41446947 on Build GRCh38 | Gain |
|
| esv3604839 | Chr.5:41252837 - 41389962 on Build GRCh38 | Gain |
|
| nsv830269 | Chr.5:41209011 - 41393434 on Build GRCh38 | Gain |
|
| nsv482997 | Chr.5:41201303 - 41350459 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map