Assay Details
Target Gene Details
Entrez Gene ID: | 60560 |
Gene Name: | N(alpha)-acetyltransferase 35, NatC auxiliary subunit |
Gene Aliases: |
EGAP, MAK10, MAK10P, bA379P1.1 |
Location: |
Chr.9:85941142-86026355 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA35 | NM_001321881.1 | NP_001308810.1 | ||
| NM_024635.3 | NP_078911.3 | |||
| XM_005252127.3 | 1 | 47 | XP_005252184.1 | |
| XM_017015012.1 | XP_016870501.1 | |||
| AK025266.1 | BAB15097.1 | |||
| AK225237.1 | ||||
| AK292239.1 | ||||
| BC117427.1 | ||||
| BC117429.1 | ||||
| DB023442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831646 | Chr.9:85760409 - 85946571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map