Assay Details
Target Gene Details
Entrez Gene ID: | 2878 |
Gene Name: | glutathione peroxidase 3 |
Gene Aliases: |
GPx-P, GSHPx-3, GSHPx-P |
Location: |
Chr.5:151020438-151028993 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPX3 | NM_002084.3 | 5 | 1294 | NP_002075.2 |
| AL036834.2 | 1 | 486 | ||
| BC013601.1 | 5 | 1124 | AAH13601.1 | |
| BC035841.1 | 5 | 1129 | AAH35841.1 | |
| BC050378.1 | 5 | 1129 | AAH50378.2 | |
| BC066949.1 | 5 | 1143 | AAH66949.1 | |
| D00632.1 | 5 | 1130 | BAA00525.1 | |
| X58295.1 | 5 | 1125 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526224 | Chr.5:151003841 - 151070675 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map