Assay Details
Target Gene Details
Entrez Gene ID: | 55743 |
Gene Name: | checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase |
Gene Aliases: |
RNF116, RNF196 |
Location: |
Chr.12:132840352-132887618 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHFR | NM_001161344.1 | 8 | 914 | NP_001154816.1 |
| NM_001161345.1 | 8 | 914 | NP_001154817.1 | |
| NM_001161346.1 | 8 | 878 | NP_001154818.1 | |
| NM_001161347.1 | 6 | 638 | NP_001154819.1 | |
| NM_018223.2 | 8 | 791 | NP_060693.2 | |
| AF170724.1 | 8 | 920 | AAF91084.1 | |
| AK001658.1 | 8 | 771 | BAA91817.1 | |
| AK027687.1 | 8 | 872 | BAB55297.1 | |
| AK097671.1 | 6 | 569 | ||
| AK302785.1 | 6 | 623 | ||
| AK304333.1 | 8 | 901 | ||
| BC012072.1 | 7 | 802 | AAH12072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561133 | Chr.12:132782284 - 132860654 on Build GRCh38 | Loss |
|
| dgv1589n100 | Chr.12:132841203 - 132967843 on Build GRCh38 | Gain |
|
| nsv455812 | Chr.12:132813412 - 132886923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map