Assay Details
Target Gene Details
Entrez Gene ID: | 100506068 |
Gene Name: | DACT3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:46660364-46677456 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DACT3-AS1 | NR_040042.1 | |||
| AW269981.1 | ||||
| AW292135.1 | ||||
| BX116982.1 |
Target Gene Details
Entrez Gene ID: | 25865 |
Gene Name: | protein kinase D2 |
Gene Aliases: |
HSPC187, PKD2, nPKC-D2 |
Location: |
Chr.19:46674316-46717127 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKD2 | NM_001079880.1 | 19 | 3200 | NP_001073349.1 |
| NM_001079881.1 | 19 | 3064 | NP_001073350.1 | |
| NM_001079882.1 | 17 | 2801 | NP_001073351.1 | |
| NM_016457.4 | 18 | 3479 | NP_057541.2 | |
| XM_005258716.2 | 17 | 2457 | XP_005258773.2 | |
| AF151021.1 | 6 | 1003 | AAF36107.1 | |
| AF309082.1 | 18 | 2761 | AAK01149.1 | |
| AK074673.1 | 17 | 2440 | BAC11127.1 | |
| AK075266.1 | 17 | 2550 | BAC11508.1 | |
| AK092896.1 | 18 | 2851 | ||
| AK095884.1 | 19 | 2743 | ||
| AK303629.1 | 17 | 2455 | ||
| AL050147.1 | 12 | 1716 | CAB43292.1 | |
| BC015472.1 | 9 | 1178 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833849 | Chr.19:46558831 - 46703060 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map