Assay Details
Target Gene Details
Entrez Gene ID: | 63035 |
Gene Name: | BCL6 corepressor-like 1 |
Gene Aliases: |
BCoR-L1, CXorf10 |
Location: |
Chr.X:129980285-130058083 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCORL1 | XM_005262452.4 | XP_005262509.2 | ||
| XM_005262453.4 | XP_005262510.2 | |||
| XM_005262454.3 | 1 | 65 | XP_005262511.2 | |
| XM_005262456.4 | XP_005262513.2 | |||
| XM_006724776.3 | XP_006724839.1 | |||
| XM_006724777.3 | XP_006724840.1 | |||
| XM_006724779.2 | 1 | 68 | XP_006724842.1 | |
| XM_017029721.1 | XP_016885210.1 | |||
| BE244710.1 | 1 | 91 | ||
| BE246581.1 | 1 | 63 | ||
| DA227697.1 | 1 | 65 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2676097 | Chr.X:129979269 - 129984916 on Build GRCh38 | Deletion |
|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map