Assay Details
Target Gene Details
Entrez Gene ID: | 64748 |
Gene Name: | phospholipid phosphatase related 2 |
Gene Aliases: |
LPPR2, PRG4 |
Location: |
Chr.19:11355386-11365698 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPPR2 | NM_001170635.1 | NP_001164106.1 | ||
| NM_022737.2 | NP_073574.2 | |||
| XM_005260017.1 | XP_005260074.1 | |||
| XM_017027147.1 | XP_016882636.1 | |||
| XM_017027148.1 | XP_016882637.1 | |||
| AK023117.1 | BAB14414.1 | |||
| AK094869.1 | BAC04443.1 | |||
| AK225482.1 | ||||
| AY304516.1 | AAP72153.1 | |||
| AY339628.1 | AAQ73540.1 | |||
| BC009378.1 | AAH09378.1 | |||
| CR936652.1 | 2 | 1476 | CAI56790.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833752 | Chr.19:11208324 - 11448939 on Build GRCh38 | Loss |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map