Assay Details
Target Gene Details
Entrez Gene ID: | 116840 |
Gene Name: | centrobin, centriole duplication and spindle assembly protein |
Gene Aliases: |
LIP8, PP1221 |
Location: |
Chr.17:7932054-7949919 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTROB | NM_001037144.5 | 1 | 249 | NP_001032221.1 |
| NM_053051.3 | 1 | 249 | NP_444279.2 | |
| XM_005256437.2 | 1 | 186 | XP_005256494.1 | |
| XM_005256438.2 | 1 | 170 | XP_005256495.1 | |
| XM_005256439.2 | 1 | 189 | XP_005256496.1 | |
| XM_017024128.1 | 1 | 186 | XP_016879617.1 | |
| XM_017024129.1 | XP_016879618.1 | |||
| XM_017024130.1 | 1 | 169 | XP_016879619.1 | |
| XM_017024131.1 | XP_016879620.1 | |||
| XM_017024132.1 | 1 | 186 | XP_016879621.1 | |
| XM_017024133.1 | XP_016879622.1 | |||
| XM_017024134.1 | 1 | 170 | XP_016879623.1 | |
| XM_017024135.1 | 1 | 155 | XP_016879624.1 | |
| XM_017024136.1 | 1 | 155 | XP_016879625.1 | |
| XM_017024137.1 | 1 | 169 | XP_016879626.1 | |
| XM_017024139.1 | 1 | 124 | XP_016879628.1 | |
| XM_017024140.1 | 1 | 151 | XP_016879629.1 | |
| XM_017024141.1 | 1 | 186 | XP_016879630.1 | |
| XM_017024142.1 | 1 | 186 | XP_016879631.1 | |
| XM_017024144.1 | 1 | 188 | XP_016879633.1 | |
| AF331638.2 | 1 | 154 | AAL56068.2 | |
| AK293076.1 | 1 | 168 | ||
| AL833907.1 | 1 | 72 | CAD38763.1 | |
| AY160226.1 | 1 | 218 | AAO22135.1 | |
| AY160227.1 | 1 | 218 | AAO22136.1 | |
| BC021134.1 | 1 | 124 | AAH21134.1 | |
| CN390730.1 | 1 | 249 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574333 | Chr.17:7880977 - 7952801 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map