Assay Details
Target Gene Details
Entrez Gene ID: | 7301 |
Gene Name: | TYRO3 protein tyrosine kinase |
Gene Aliases: |
BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif |
Location: |
Chr.15:41559022-41579338 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TYRO3 | NM_006293.3 | NP_006284.2 | ||
| XM_017022542.1 | XP_016878031.1 | |||
| XM_017022543.1 | XP_016878032.1 | |||
| AB209431.1 | BAD92668.1 | |||
| AK295496.1 | ||||
| BC029925.2 | AAH29925.2 | |||
| BC049368.1 | AAH49368.1 | |||
| BC051756.1 | AAH51756.1 | |||
| D17517.1 | BAA04467.1 | |||
| D50479.1 | BAA21781.1 | |||
| U02566.1 | AAA17407.1 | |||
| U05682.1 | AAA19236.1 | |||
| X72886.1 | CAA51396.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3552368 | Chr.15:41573108 - 41577990 on Build GRCh38 | Deletion |
|
| esv2749620 | Chr.15:41573479 - 41577890 on Build GRCh38 | Deletion |
|
| dgv1200n106 | Chr.15:41573466 - 41573712 on Build GRCh38 | Deletion |
|
| nsv1069854 | Chr.15:41573451 - 41573675 on Build GRCh38 | Deletion |
|
| esv2749618 | Chr.15:41573459 - 41573684 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map