Assay Details
Target Gene Details
Entrez Gene ID: | 78987 |
Gene Name: | cysteine rich with EGF like domains 1 |
Gene Aliases: |
AVSD2, CIRRIN |
Location: |
Chr.3:9933788-9945413 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRELD1 | NM_001031717.3 | NP_001026887.1 | ||
| NM_001077415.2 | NP_001070883.1 | |||
| NM_015513.4 | NP_056328.2 | |||
| XM_011534108.1 | XP_011532410.1 | |||
| XM_017007175.1 | XP_016862664.1 | |||
| AF452623.1 | AAM75206.1 | |||
| AK314113.1 | ||||
| AL050275.1 | CAB43376.1 | |||
| AY358363.1 | AAQ88729.1 | |||
| BC008720.2 | AAH08720.1 | |||
| BX358821.2 | ||||
| CR457380.1 | CAG33661.1 | |||
| DN996843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss |
|
| nsv589648 | Chr.3:9892921 - 10042251 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map