Assay Details
Target Gene Details
Entrez Gene ID: | 9255 |
Gene Name: | aminoacyl tRNA synthetase complex interacting multifunctional protein 1 |
Gene Aliases: |
EMAP2, EMAPII, HLD3, SCYE1, p43 |
Location: |
Chr.4:106315610-106349225 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIMP1 | NM_001142415.1 | NP_001135887.1 | ||
| NM_004757.3 | 1 | 470 | NP_004748.2 | |
| AK095951.1 | 1 | 470 | ||
| AK297950.1 | ||||
| BG941396.1 | 1 | 78 | ||
| DA211424.1 | 1 | 71 | ||
| DC358150.1 |
Target Gene Details
Entrez Gene ID: | 93627 |
Gene Name: | TBC1 domain containing kinase |
Gene Aliases: |
HSPC302, IHPRF3, TBCKL |
Location: |
Chr.4:106044317-106316704 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCK | NM_001163435.2 | 1 | 102 | NP_001156907.1 |
| NM_001163436.2 | NP_001156908.1 | |||
| NM_001163437.2 | 1 | 102 | NP_001156909.1 | |
| NM_001290768.1 | 1 | 102 | NP_001277697.1 | |
| NM_033115.4 | 1 | 102 | NP_149106.2 | |
| XM_017008846.1 | 1 | 102 | XP_016864335.1 | |
| AK074305.1 | BAB85045.1 | |||
| AK315571.1 | 1 | 45 | ||
| BC009208.2 | 1 | 64 | AAH09208.2 | |
| BC068496.1 | 1 | 43 | AAH68496.1 | |
| BX647851.1 | 1 | 46 | CAI46102.1 | |
| DA100605.1 | 1 | 102 | ||
| DB223368.1 | ||||
| DB228807.1 | 1 | 102 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830029 | Chr.4:106281939 - 106432326 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map