Assay Details
Target Gene Details
Entrez Gene ID: | 51318 |
Gene Name: | mitochondrial ribosomal protein L35 |
Gene Aliases: |
L35mt, MRP-L35 |
Location: |
Chr.2:86199355-86213794 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL35 | NM_016622.3 | 4 | 1054 | NP_057706.2 |
| NM_145644.2 | NP_663619.1 | |||
| XM_005264361.2 | XP_005264418.1 | |||
| AK074352.1 | 4 | 1026 | ||
| AL832982.1 | 4 | 1002 | CAH56349.1 | |
| BC020651.1 | AAH20651.1 | |||
| BQ421154.1 | ||||
| BQ879834.1 | 1 | 49 | ||
| DB245340.1 | 1 | 236 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458563 | Chr.2:86065218 - 86280565 on Build GRCh38 | Gain |
|
| dgv3885n100 | Chr.2:86054844 - 86279009 on Build GRCh38 | Gain |
|
| nsv582372 | Chr.2:86158605 - 86280565 on Build GRCh38 | Gain |
|
| esv2751903 | Chr.2:86026419 - 86278330 on Build GRCh38 | Gain |
|
| dgv713e214 | Chr.2:86173703 - 86286364 on Build GRCh38 | Gain |
|
| esv3591540 | Chr.2:86055590 - 86283809 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map