Assay Details
Target Gene Details
Entrez Gene ID: | 79858 |
Gene Name: | NIMA related kinase 11 |
Gene Aliases: |
- |
Location: |
Chr.3:131026850-131350465 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
More Information
Set Membership: |
|

Genomic Map