Assay Details
Target Gene Details
Entrez Gene ID: | 114609 |
Gene Name: | TIR domain containing adaptor protein |
Gene Aliases: |
BACTS1, Mal, MyD88-2, wyatt |
Location: |
Chr.11:126281268-126294933 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIRAP | NM_001039661.1 | NP_001034750.1 | ||
| NM_001318776.1 | NP_001305705.1 | |||
| NM_001318777.1 | NP_001305706.1 | |||
| NM_148910.2 | 2 | 189 | NP_683708.1 | |
| XM_005271399.3 | XP_005271456.2 | |||
| XM_011542576.2 | XP_011540878.1 | |||
| XM_017017161.1 | XP_016872650.1 | |||
| XM_017017162.1 | 2 | 210 | XP_016872651.1 | |
| AF378129.1 | 2 | 170 | AAL05627.1 | |
| AF410783.1 | 2 | 166 | AAL05036.1 | |
| AK124298.1 | ||||
| AK313147.1 | ||||
| BC032474.1 | AAH32474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map