Assay Details
Target Gene Details
Entrez Gene ID: | 147906 |
Gene Name: | dishevelled binding antagonist of beta catenin 3 |
Gene Aliases: |
DAPPER3, RRR1 |
Location: |
Chr.19:46647564-46661138 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DACT3 | NM_001301046.1 | 3 | 2066 | NP_001287975.1 |
| NM_145056.2 | 4 | 2374 | NP_659493.2 | |
| XM_011526499.2 | 4 | 3150 | XP_011524801.1 | |
| XM_011526500.2 | 3 | 2066 | XP_011524802.1 | |
| XM_017026334.1 | 4 | 2235 | XP_016881823.1 | |
| AI769079.1 | ||||
| BC016161.1 | 3 | 2066 | AAH16161.1 | |
| BX097198.1 | 1 | 323 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828570 | Chr.19:46637434 - 46652691 on Build GRCh38 | Gain |
|
| nsv833849 | Chr.19:46558831 - 46703060 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map