Assay Details
Target Gene Details
Entrez Gene ID: | 7033 |
Gene Name: | trefoil factor 3 |
Gene Aliases: |
ITF, P1B, TFI |
Location: |
Chr.21:42311667-42315596 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFF3 | NM_003226.3 | NP_003217.3 | ||
| AF432265.1 | AAL28111.1 | |||
| BC017859.1 | AAH17859.1 | |||
| BQ016069.1 | ||||
| L08044.1 | AAA36766.1 | |||
| L15203.1 | AAA59981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510799 | Chr.21:42255022 - 42324525 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map