Assay Details
Target Gene Details
Entrez Gene ID: | 643866 |
Gene Name: | cerebellin 3 precursor |
Gene Aliases: |
PRO1486 |
Location: |
Chr.14:24426532-24429525 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBLN3 | NM_001039771.2 | 1 | 255 | NP_001034860.1 |
| AY359070.1 | 1 | 255 | AAQ89429.1 |
Target Gene Details
Entrez Gene ID: | 23351 |
Gene Name: | KH and NYN domain containing |
Gene Aliases: |
KIAA0323 |
Location: |
Chr.14:24423186-24441342 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KHNYN | NM_001290256.1 | NP_001277185.1 | ||
| XM_011536590.2 | XP_011534892.1 | |||
| AB002321.1 |
More Information
Set Membership: |
|

Genomic Map