Assay Details
Target Gene Details
Entrez Gene ID: | 9962 |
Gene Name: | solute carrier family 23 member 2 |
Gene Aliases: |
NBTL1, SLC23A1, SVCT2, YSPL2, hSVCT2 |
Location: |
Chr.20:4852356-5010293 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC23A2 | NM_005116.5 | 17 | 4717 | NP_005107.4 |
| NM_203327.1 | 17 | 4735 | NP_976072.1 | |
| XM_011529414.2 | 17 | 4722 | XP_011527716.1 | |
| XM_011529415.2 | 16 | 4543 | XP_011527717.1 | |
| XM_011529416.2 | 16 | 4852 | XP_011527718.1 | |
| XM_011529417.2 | 16 | 4599 | XP_011527719.1 | |
| XM_017028171.1 | 17 | 4756 | XP_016883660.1 | |
| XM_017028172.1 | 17 | 4760 | XP_016883661.1 | |
| XM_017028173.1 | 17 | 4740 | XP_016883662.1 | |
| XM_017028174.1 | 17 | 4635 | XP_016883663.1 | |
| XM_017028175.1 | 13 | 4071 | XP_016883664.1 | |
| XM_017028176.1 | 14 | 4389 | XP_016883665.1 | |
| XM_017028177.1 | 14 | 4376 | XP_016883666.1 | |
| XM_017028178.1 | 16 | 4642 | XP_016883667.1 | |
| XM_017028179.1 | 16 | 4629 | XP_016883668.1 | |
| XM_017028180.1 | 12 | 3953 | XP_016883669.1 | |
| AK025668.1 | 1 | 1521 | ||
| AK297253.1 | ||||
| D87075.2 | 17 | 4717 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645074 | Chr.20:4205961 - 5037636 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map