Assay Details
Target Gene Details
Entrez Gene ID: | 162 |
Gene Name: | adaptor related protein complex 1 beta 1 subunit |
Gene Aliases: |
ADTB1, AP105A, BAM22, CLAPB2 |
Location: |
Chr.22:29327680-29388583 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1B1 | NM_001127.3 | 23 | 3654 | NP_001118.3 |
| NM_001166019.1 | 21 | 3564 | NP_001159491.1 | |
| NM_145730.2 | 21 | 3624 | NP_663782.2 | |
| AK294406.1 | 14 | 2218 | ||
| BC046242.1 | 21 | 3564 | AAH46242.1 | |
| BX537680.1 | 4 | 3036 | CAD97809.1 | |
| CD364485.1 | 1 | 541 | ||
| L13939.1 | 22 | 3513 | AAC98702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834171 | Chr.22:29230413 - 29433855 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116328912] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map