Assay Details
Target Gene Details
Entrez Gene ID: | 9743 |
Gene Name: | Rho GTPase activating protein 32 |
Gene Aliases: |
GC-GAP, GRIT, PX-RICS, RICS, p200RhoGAP, p250GAP |
Location: |
Chr.11:128965060-129279168 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP32 | NM_001142685.1 | 21 | 3123 | NP_001136157.1 |
| NM_014715.3 | 12 | 2455 | NP_055530.2 | |
| XM_005271736.4 | 22 | 4153 | XP_005271793.2 | |
| XM_011543072.2 | 22 | 3355 | XP_011541374.1 | |
| XM_011543073.2 | 22 | 3091 | XP_011541375.2 | |
| XM_011543075.2 | 13 | 2301 | XP_011541377.1 | |
| XM_017018595.1 | 22 | 3072 | XP_016874084.1 | |
| XM_017018596.1 | 20 | 3349 | XP_016874085.1 | |
| AB018255.2 | 12 | 2376 | ||
| AB088416.1 | 12 | 2376 | ||
| AL833062.2 | 19 | 3616 | CAD89974.1 | |
| AY194287.1 | 12 | 2379 | AAO43677.1 | |
| BC000277.1 | AAH00277.2 | |||
| BC104898.1 | 11 | 2110 | AAI04899.1 | |
| BC113429.1 | 11 | 2110 | ||
| EF127492.1 | 21 | 3123 |
More Information
Set Membership: |
|

Genomic Map