Assay Details
Target Gene Details
Entrez Gene ID: | 29960 |
Gene Name: | mitochondrial rRNA methyltransferase 2 |
Gene Aliases: |
FJH1, FTSJ2, HEL97, RRMJ2 |
Location: |
Chr.7:2234291-2242198 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRM2 | NM_013393.1 | NP_037525.1 | ||
| AF093415.1 | AAF22488.1 | |||
| AK302060.1 | ||||
| BC114514.1 | AAI14515.1 | |||
| BC114564.1 | AAI14565.1 | |||
| CR749618.1 | CAH18412.1 | |||
| EU794658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021310 | Chr.7:1936627 - 2280155 on Build GRCh38 | Loss |
|
| nsv1018563 | Chr.7:2233301 - 2325297 on Build GRCh38 | Gain |
|
| nsv1021173 | Chr.7:2206456 - 2334992 on Build GRCh38 | Loss |
|
| nsv830886 | Chr.7:2145358 - 2337903 on Build GRCh38 | Loss |
|
| esv2761136 | Chr.7:2230260 - 2325309 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map