Assay Details
Target Gene Details
Entrez Gene ID: | 6947 |
Gene Name: | transcobalamin 1 |
Gene Aliases: |
HC, TC-1, TC1, TCI |
Location: |
Chr.11:59852808-59866568 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCN1 | NM_001062.3 | 3 | 413 | NP_001053.2 |
| AK292990.1 | 3 | 330 | ||
| BP370727.1 | 3 | 419 | ||
| J05068.1 | 3 | 390 | AAA61058.1 |
More Information
Set Membership: |
|

Genomic Map