Assay Details
Target Gene Details
Entrez Gene ID: | 3559 |
Gene Name: | interleukin 2 receptor subunit alpha |
Gene Aliases: |
CD25, IDDM10, IL2R, IMD41, TCGFR, p55 |
Location: |
Chr.10:6010694-6062370 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL2RA | NM_000417.2 | 8 | 1229 | NP_000408.1 |
| NM_001308242.1 | 7 | 1013 | NP_001295171.1 | |
| NM_001308243.1 | 6 | 941 | NP_001295172.1 | |
| AK223313.1 | 8 | 1151 | BAD97033.1 | |
| CR996090.1 | ||||
| K03122.1 | 7 | 953 | AAB59535.1 | |
| X01057.1 | 8 | 1190 | CAA25525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map