Assay Details
Target Gene Details
Entrez Gene ID: | 116448 |
Gene Name: | oligodendrocyte transcription factor 1 |
Gene Aliases: |
BHLHB6, BHLHE21 |
Location: |
Chr.21:33070144-33072422 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLIG1 | NM_138983.2 | 1 | 2105 | NP_620450.2 |
| AL355743.1 | 1 | 1249 | ||
| AL834450.1 | 1 | 2098 | CAD39110.1 | |
| AW298788.1 | 1 | 189 | ||
| BC026989.1 | 1 | 1982 | AAH26989.1 | |
| BC033290.1 | 1 | 2112 | AAH33290.1 |
More Information
Set Membership: |
|

Genomic Map