Assay Details
Target Gene Details
Entrez Gene ID: | 2206 |
Gene Name: | membrane spanning 4-domains A2 |
Gene Aliases: |
APY, ATOPY, FCER1B, FCERI, IGEL, IGER, IGHER, MS4A1 |
Location: |
Chr.11:60088261-60098467 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A2 | NM_000139.4 | 5 | 592 | NP_000130.1 |
| NM_001256916.1 | 4 | 457 | NP_001243845.1 | |
| XM_005273846.4 | 6 | 907 | XP_005273903.1 | |
| XM_011544850.2 | 6 | 525 | XP_011543152.1 | |
| XM_017017362.1 | 6 | 907 | XP_016872851.1 | |
| AK304487.1 | 5 | 592 | ||
| AK310142.1 | 5 | 592 | ||
| BC074800.2 | 5 | 521 | AAH74800.1 | |
| BC074843.2 | 5 | 521 | AAH74843.1 | |
| D10583.1 | 5 | 540 | BAA01440.1 | |
| DA972757.1 | ||||
| JF411082.1 | 4 | 355 | ||
| X66187.1 | 3 | 280 | CAA46955.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468579 | Chr.11:60046814 - 60207927 on Build GRCh38 | Gain |
|
| dgv201e214 | Chr.11:60084926 - 60104676 on Build GRCh38 | Loss |
|
| esv3626549 | Chr.11:60093431 - 60115836 on Build GRCh38 | Loss |
|
| nsv832173 | Chr.11:60021411 - 60173087 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map