Assay Details
Target Gene Details
Entrez Gene ID: | 256691 |
Gene Name: | MAM domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.9:70043519-70226972 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAMDC2 | NM_153267.4 | 14 | 3295 | NP_694999.3 |
| NR_125850.1 | 14 | 3291 | ||
| BC015417.2 | 6 | 1359 | AAH15417.1 | |
| BC016383.1 | 14 | 3013 | AAH16383.1 | |
| BC063634.1 | 14 | 3272 | AAH63634.1 | |
| BX538309.1 | 3 | 4286 | CAD98089.1 |
Target Gene Details
Entrez Gene ID: | 100507299 |
Gene Name: | SMC5 antisense RNA 1 (head to head) |
Gene Aliases: |
MAMDC2-AS2 |
Location: |
Chr.9:70216059-70258874 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMC5-AS1 | NR_039990.1 | |||
| AK124136.1 | ||||
| CR936772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6555 | Chr.9:70225054 - 70259811 on Build GRCh38 | Insertion |
|
| nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain |
|

Genomic Map