Assay Details
Target Gene Details
Entrez Gene ID: | 149699 |
Gene Name: | gametocyte specific factor 1 like |
Gene Aliases: |
C20orf65, FAM112A, dJ1028D15.4 |
Location: |
Chr.20:43726161-43727135 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTSF1L | NM_001008901.1 | 1 | 302 | NP_001008901.1 |
| NM_176791.3 | 1 | 302 | NP_789761.1 | |
| XM_005260298.4 | 1 | 435 | XP_005260355.1 | |
| AA813527.1 | ||||
| AK058060.1 | 1 | 298 | ||
| BC040049.1 | 1 | 227 | AAH40049.1 | |
| BQ428678.1 | 1 | 306 | ||
| BX355734.2 | 1 | 222 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645889 | Chr.20:43686024 - 43756429 on Build GRCh38 | Gain |
|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| esv3645891 | Chr.20:43718815 - 43731133 on Build GRCh38 | Loss |
|
| esv3645886 | Chr.20:43653014 - 43756886 on Build GRCh38 | Gain |
|
| esv2673612 | Chr.20:43719016 - 43731241 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map