Assay Details
Target Gene Details
Entrez Gene ID: | 1879 |
Gene Name: | early B-cell factor 1 |
Gene Aliases: |
COE1, EBF, O/E-1, OLF1 |
Location: |
Chr.5:158695915-159099762 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBF1 | NM_001290360.2 | 16 | 2593 | NP_001277289.1 |
| NM_001324101.1 | 17 | 2662 | NP_001311030.1 | |
| NM_001324103.1 | 17 | 2659 | NP_001311032.1 | |
| NM_001324106.1 | 17 | 2635 | NP_001311035.1 | |
| NM_001324107.1 | 16 | 2569 | NP_001311036.1 | |
| NM_001324108.1 | 16 | 2566 | NP_001311037.1 | |
| NM_001324109.1 | 16 | 2482 | NP_001311038.1 | |
| NM_001324111.1 | 16 | 2688 | NP_001311040.1 | |
| NM_024007.4 | 16 | 2590 | NP_076870.1 | |
| NM_182708.2 | 15 | 2497 | NP_874367.1 | |
| XM_017009198.1 | 16 | 2950 | XP_016864687.1 | |
| XM_017009201.1 | 11 | 20077 | XP_016864690.1 | |
| AK123757.1 | 1 | 181 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5093 | Chr.5:158691822 - 158725743 on Build GRCh38 | Insertion |
|
| nsv830528 | Chr.5:158698214 - 158913021 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map