Assay Details
Target Gene Details
Entrez Gene ID: | 541468 |
Gene Name: | leucine rich adaptor protein 1 |
Gene Aliases: |
C1orf190, LRAP35a, LRP35A |
Location: |
Chr.1:46203334-46221256 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LURAP1 | NM_001013615.2 | 2 | 349 | NP_001013633.1 |
| AK057892.1 | 2 | 349 | BAB71608.1 | |
| BC034422.1 | 2 | 342 | AAH34422.1 |
Target Gene Details
Entrez Gene ID: | 55624 |
Gene Name: | protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-) |
Gene Aliases: |
GNTI.2, GnT I.2, LGMD2O, MEB, MGAT1.2, gnT-I.2 |
Location: |
Chr.1:46188681-46220305 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POMGNT1 | NM_001243766.1 | 1 | 550 | NP_001230695.1 |
| AK056186.1 | 1 | 550 | ||
| CR749700.1 | 1 | 536 | CAH18478.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470711 | Chr.1:46194623 - 46344858 on Build GRCh38 | Gain |
|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map