Assay Details
Target Gene Details
Entrez Gene ID: | 80310 |
Gene Name: | platelet derived growth factor D |
Gene Aliases: |
IEGF, MSTP036, SCDGF-B, SCDGFB |
Location: |
Chr.11:103907186-104164299 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFD | NM_025208.4 | 7 | 1442 | NP_079484.1 |
| NM_033135.3 | 7 | 1424 | NP_149126.1 | |
| AB033832.1 | 7 | 1226 | BAB18903.1 | |
| AF113216.1 | 7 | 1176 | AAG39287.1 | |
| AF335584.1 | 7 | 1251 | AAK38840.1 | |
| AF336376.1 | 7 | 1245 | AAK56136.1 | |
| AK292801.1 | 7 | 1250 | ||
| AY027517.1 | 7 | 1162 | AAK20081.1 | |
| AY027518.1 | 7 | 1144 | AAK20082.1 | |
| AY359116.1 | 7 | 1083 | AAQ89474.1 | |
| BC030645.2 | 7 | 1504 | AAH30645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750977 | Chr.11:103211829 - 104054062 on Build GRCh38 | Gain |
|
| esv2760322 | Chr.11:103889357 - 104503413 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map