Assay Details
Target Gene Details
Entrez Gene ID: | 3976 |
Gene Name: | leukemia inhibitory factor |
Gene Aliases: |
CDF, DIA, HILDA, MLPLI |
Location: |
Chr.22:30240447-30246851 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIF | NM_001257135.1 | 2 | 1778 | NP_001244064.1 |
| NM_002309.4 | 3 | 1957 | NP_002300.1 | |
| X13967.1 | 3 | 1845 | CAA32147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834173 | Chr.22:30119103 - 30262288 on Build GRCh38 | Loss |
|
| nsv834174 | Chr.22:30223771 - 30350926 on Build GRCh38 | Gain+Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73881421] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map