Assay Details
Target Gene Details
Entrez Gene ID: | 10148 |
Gene Name: | Epstein-Barr virus induced 3 |
Gene Aliases: |
IL-27B, IL27B |
Location: |
Chr.19:4229196-4237539 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBI3 | NM_005755.2 | 5 | 696 | NP_005746.2 |
| XM_011527619.2 | 6 | 945 | XP_011525921.1 | |
| BC015364.1 | 5 | 741 | AAH15364.1 | |
| BC046112.1 | 5 | 695 | AAH46112.1 | |
| L08187.1 | 5 | 701 | AAA93193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578442 | Chr.19:4236999 - 4312106 on Build GRCh38 | Loss |
|
| nsv833717 | Chr.19:4204757 - 4368519 on Build GRCh38 | Loss |
|
| nsv833718 | Chr.19:4221577 - 4480248 on Build GRCh38 | Loss |
|
| nsv833716 | Chr.19:4121259 - 4274854 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map