Assay Details
Target Gene Details
Entrez Gene ID: | 58504 |
Gene Name: | Rho GTPase activating protein 22 |
Gene Aliases: |
RhoGAP2, RhoGap22 |
Location: |
Chr.10:48429831-48656265 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950952 | Chr.10:48445358 - 48458757 on Build GRCh38 | Deletion |
|
| nsv1039508 | Chr.10:48387675 - 48446360 on Build GRCh38 | Gain |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map