Assay Details
Target Gene Details
Entrez Gene ID: | 83591 |
Gene Name: | THAP domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.12:71663897-71680648 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THAP2 | NM_031435.3 | 3 | 3523 | NP_113623.1 |
| AL136607.1 | 3 | 3420 | CAB66542.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832451 | Chr.12:71584552 - 71806081 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map