Assay Details
Target Gene Details
Entrez Gene ID: | 23092 |
Gene Name: | Rho GTPase activating protein 26 |
Gene Aliases: |
GRAF, GRAF1, OPHN1L, OPHN1L1 |
Location: |
Chr.5:142770370-143229007 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP26 | NM_001135608.1 | 1 | 83 | NP_001129080.1 |
| NM_015071.4 | 1 | 83 | NP_055886.1 | |
| XM_005268402.4 | 1 | 437 | XP_005268459.1 | |
| BC068555.1 | 1 | 403 | AAH68555.1 | |
| KU178561.1 | 1 | 48 | ||
| KU178562.1 | 1 | 48 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606999 | Chr.5:142429551 - 143065624 on Build GRCh38 | Gain |
|
| nsv830510 | Chr.5:142623697 - 142804053 on Build GRCh38 | Gain |
|
| nsv1031126 | Chr.5:142514198 - 143020006 on Build GRCh38 | Gain |
|
| esv3607005 | Chr.5:142613395 - 142792182 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map