Assay Details
Target Gene Details
Entrez Gene ID: | 50848 |
Gene Name: | F11 receptor |
Gene Aliases: |
CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1 |
Location: |
Chr.1:160995211-161021343 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F11R | NM_016946.4 | 10 | 1446 | NP_058642.1 |
| AF154005.1 | 10 | 1118 | AAD43794.1 | |
| AF172398.2 | 10 | 1322 | AAD48877.1 | |
| AF207907.1 | 10 | 1191 | AAF22829.1 | |
| AK026665.1 | 10 | 1237 | ||
| AK075152.1 | 10 | 1246 | BAC11436.1 | |
| AK304412.1 | 9 | 1052 | ||
| AL136649.1 | 10 | 1250 | CAB66584.1 | |
| AY358896.1 | 10 | 1227 | AAQ89255.1 | |
| BC001533.2 | 10 | 1220 | AAH01533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548058 | Chr.1:160955665 - 161019824 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map