Assay Details
Target Gene Details
Entrez Gene ID: | 1103 |
Gene Name: | choline O-acetyltransferase |
Gene Aliases: |
CHOACTASE, CMS1A, CMS1A2, CMS6 |
Location: |
Chr.10:49609095-49665104 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHAT | NM_001142929.1 | NP_001136401.1 | ||
| NM_001142933.1 | NP_001136405.1 | |||
| NM_001142934.1 | NP_001136406.1 | |||
| NM_020549.4 | NP_065574.3 | |||
| NM_020984.3 | NP_066264.3 | |||
| NM_020985.3 | NP_066265.3 | |||
| NM_020986.3 | NP_066266.3 | |||
| AF305907.1 | AAK08953.1 | |||
| AF305908.2 | AAK08954.1 | |||
| AF305909.1 | AAK08955.1 | |||
| D82339.1 | BAA18943.1 | |||
| D82340.1 | BAA18944.1 | |||
| D82341.1 | BAA18945.1 | |||
| D82342.1 | BAA18946.1 | |||
| S56138.1 | AAA14245.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950954 | Chr.10:49609255 - 49617854 on Build GRCh38 | Deletion |
|
| nsv550819 | Chr.10:49607466 - 49627218 on Build GRCh38 | Loss |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map