Assay Details
Target Gene Details
Entrez Gene ID: | 26040 |
Gene Name: | SET binding protein 1 |
Gene Aliases: |
MRD29, SEB |
Location: |
Chr.18:44680173-45068510 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETBP1 | NM_001130110.1 | 2 | 567 | NP_001123582.1 |
| NM_015559.2 | 2 | 468 | NP_056374.2 | |
| AB007897.2 | 2 | 468 | ||
| AB022660.1 | BAA82444.1 | |||
| BC062338.1 | 2 | 567 | AAH62338.1 | |
| BC146776.1 | 2 | 468 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2272 | Chr.18:44678516 - 44723338 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map