Assay Details
Target Gene Details
Entrez Gene ID: | 9442 |
Gene Name: | mediator complex subunit 27 |
Gene Aliases: |
CRAP34, CRSP34, CRSP8, MED3, TRAP37 |
Location: |
Chr.9:131860110-132079887 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED27 | NM_001253881.1 | NP_001240810.1 | ||
| NM_004269.3 | NP_004260.2 | |||
| XM_017015329.1 | XP_016870818.1 | |||
| AF104252.1 | AAD12721.1 | |||
| AF230382.1 | AAF37290.1 | |||
| AK124232.1 | ||||
| BC002878.1 | AAH02878.1 | |||
| BC064608.1 | AAH64608.1 | |||
| DQ099387.1 | AAZ13763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621898 | Chr.9:131733919 - 131887845 on Build GRCh38 | Gain |
|
| esv3621902 | Chr.9:131800850 - 131870335 on Build GRCh38 | Gain |
|
| nsv1038424 | Chr.9:131788536 - 131906562 on Build GRCh38 | Gain |
|
| nsv1051484 | Chr.9:131850783 - 131872374 on Build GRCh38 | Loss |
|
| esv3621896 | Chr.9:131651740 - 131887259 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map