Assay Details
Target Gene Details
Entrez Gene ID: | 221264 |
Gene Name: | adenylate kinase 9 |
Gene Aliases: |
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1 |
Location: |
Chr.6:109492855-109691253 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 47 - Exon 47 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK9 | NM_001145128.2 | NP_001138600.2 | ||
| XM_006715376.3 | XP_006715439.2 | |||
| XM_011535550.2 | XP_011533852.1 | |||
| XM_011535552.2 | XP_011533854.1 | |||
| XM_011535553.2 | XP_011533855.1 | |||
| XM_011535554.2 | XP_011533856.1 | |||
| XM_011535558.1 | XP_011533860.1 | |||
| XM_017010385.1 | XP_016865874.1 | |||
| XM_017010387.1 | XP_016865876.1 | |||
| XM_017010389.1 | XP_016865878.1 | |||
| AK098657.1 | BAC05368.1 | |||
| BC101714.1 | AAI01715.1 | |||
| BC111948.1 | AAI11949.1 | |||
| BC143533.1 |
More Information
Set Membership: |
|

Genomic Map