Assay Details
Target Gene Details
Entrez Gene ID: | 125150 |
Gene Name: | zinc finger SWIM-type containing 7 |
Gene Aliases: |
SWS1 |
Location: |
Chr.17:15976560-15999692 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSWIM7 | NM_001042697.1 | NP_001036162.1 | ||
| NM_001042698.1 | NP_001036163.1 | |||
| AK093384.1 | ||||
| BU075283.1 | ||||
| DQ530600.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3108n100 | Chr.17:15966243 - 16193927 on Build GRCh38 | Gain |
|
| esv3892973 | Chr.17:15947882 - 16030711 on Build GRCh38 | Gain |
|
| esv3572431 | Chr.17:15951820 - 16194089 on Build GRCh38 | Gain |
|
| dgv3107n100 | Chr.17:15703953 - 16150198 on Build GRCh38 | Gain |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map