Assay Details
Target Gene Details
Entrez Gene ID: | 84148 |
Gene Name: | lysine acetyltransferase 8 |
Gene Aliases: |
MOF, MYST1, ZC2HC8, hMOF |
Location: |
Chr.16:31117664-31131393 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT8 | NM_032188.2 | 5 | 585 | NP_115564.2 |
| NM_182958.2 | 5 | 585 | NP_892003.2 | |
| XM_011545970.1 | 6 | 1126 | XP_011544272.1 | |
| XM_011545971.1 | 6 | 1131 | XP_011544273.1 | |
| AF217501.1 | 5 | 551 | AAL56648.1 | |
| AF260665.2 | 5 | 483 | AAF72665.2 | |
| AF289578.1 | 2 | 1513 | AAL55762.1 | |
| AK021872.1 | 5 | 582 | BAB13924.1 | |
| AK024102.1 | 5 | 583 | BAB14827.1 | |
| AK225660.1 | 5 | 585 | ||
| AK291106.1 | 5 | 582 | ||
| AL050395.1 | CAH56416.1 | |||
| BC037773.1 | 5 | 582 | AAH37773.1 | |
| BP342476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571820 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map