Assay Details
Target Gene Details
Entrez Gene ID: | 25819 |
Gene Name: | nocturnin |
Gene Aliases: |
CCR4L, CCRN4L, Ccr4c, NOC |
Location: |
Chr.4:139015759-139045939 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOCT | NM_012118.3 | NP_036250.2 | ||
| AF183961.1 | AAD56548.1 | |||
| AF199493.1 | AAG01388.1 | |||
| BC021963.2 | 2 | 636 | AAH21963.1 | |
| BC023512.2 | 2 | 636 | AAH23512.1 | |
| BC077723.1 | 2 | 636 | AAH77723.1 | |
| BC113494.1 | ||||
| BC113500.1 |
More Information
Set Membership: |
|

Genomic Map