Assay Details
Target Gene Details
Entrez Gene ID: | 79607 |
Gene Name: | family with sequence similarity 118 member B |
Gene Aliases: |
- |
Location: |
Chr.11:126211414-126264540 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM118B | XM_011542977.2 | 8 | 3459 | XP_011541279.1 |
Target Gene Details
Entrez Gene ID: | 6734 |
Gene Name: | SRP receptor alpha subunit |
Gene Aliases: |
DP, SRPR, Sralpha |
Location: |
Chr.11:126255151-126268982 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPRA | NM_001177842.1 | 11 | 1737 | NP_001171313.1 |
| NM_003139.3 | 12 | 1821 | NP_003130.2 | |
| XM_017018179.1 | 12 | 1767 | XP_016873668.1 | |
| AK130382.1 | 9 | 1236 | ||
| AK303379.1 | 11 | 1737 | ||
| AK312377.1 | 12 | 1739 | ||
| BC001162.1 | 12 | 1701 | AAH01162.1 | |
| BC009110.1 | 12 | 1664 | AAH09110.1 | |
| BC013583.1 | 12 | 1727 | AAH13583.1 | |
| CR457025.1 | 12 | 1643 | CAG33306.1 | |
| X06272.1 | 12 | 1676 | CAA29608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556529 | Chr.11:126204297 - 126264724 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map