Assay Details
Target Gene Details
Entrez Gene ID: | 10608 |
Gene Name: | MAX dimerization protein 4 |
Gene Aliases: |
MAD4, MST149, MSTP149, bHLHc12 |
Location: |
Chr.4:2247432-2262143 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MXD4 | NM_006454.2 | NP_006445.1 | ||
| XM_011513388.2 | XP_011511690.1 | |||
| AF040963.1 | AAB97009.1 | |||
| AF082740.1 | AAF98553.1 | |||
| AF187724.1 | AAQ13695.1 | |||
| AK024501.1 | BAB15791.1 | |||
| BC002713.2 | AAH02713.1 | |||
| BC068060.1 | AAH68060.1 | |||
| BT019969.1 | AAV38772.1 | |||
| BT019970.1 | AAV38773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508990 | Chr.4:2246239 - 2324327 on Build GRCh38 | Insertion |
|
| nsv593377 | Chr.4:2246106 - 2261919 on Build GRCh38 | Loss |
|
| nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| dgv312n21 | Chr.4:2243138 - 2305739 on Build GRCh38 | Loss |
|
| dgv8910n54 | Chr.4:2249015 - 2262496 on Build GRCh38 | Loss |
|
| nsv829838 | Chr.4:2155167 - 2349950 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map