Assay Details
Target Gene Details
Entrez Gene ID: | 2043 |
Gene Name: | EPH receptor A4 |
Gene Aliases: |
HEK8, SEK, TYRO1 |
Location: |
Chr.2:221418027-221574202 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA4 | NM_001304536.1 | 19 | 5766 | NP_001291465.1 |
| NM_001304537.1 | 17 | 5533 | NP_001291466.1 | |
| NM_004438.4 | 18 | 5601 | NP_004429.1 | |
| XM_005246374.2 | 18 | 5630 | XP_005246431.1 | |
| AA987900.1 | 1 | 62 | ||
| AK225181.1 | 4 | 835 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584512 | Chr.2:221255109 - 222040142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map